ENA394768162 Enamine

2-(3,5-dimethylphenyl)-2H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD16699916
  • Cas Number : 1249437-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129679
Availability details for
Quantity
  • Availability
  • Add to Cart