ENA394768163 Enamine

N-(1-phenylethyl)hydroxylamine

  • MDL number MFCD06660399
  • Cas Number : 2912-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129680
Availability details for
Quantity
  • Availability
  • Add to Cart