ENA394768167 Enamine

2-(3,5-dimethoxyphenyl)-2-hydroxyacetonitrile

  • MDL number MFCD12169290
  • Cas Number : 160708-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130322

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130322
Availability details for
Quantity
  • Availability
  • Add to Cart