ENA394768172 Enamine

1-(4-chloro-2-fluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD17290246
  • Cas Number : 1341892-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130333
Availability details for
Quantity
  • Availability
  • Add to Cart