ENA394768176 Enamine

1-(bromomethyl)-5-methoxy-4-(2-methylbutoxy)-2-nitrobenzene

  • MDL number MFCD17301546
  • Cas Number : 1275548-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130338
Availability details for
Quantity
  • Availability
  • Add to Cart