ENA394768178 Enamine

methyl 3-(4-bromobutoxy)benzoate

  • MDL number MFCD12192961
  • Cas Number : 486396-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130341
Availability details for
Quantity
  • Availability
  • Add to Cart