ENA394768179 Enamine

methyl 4-(4-bromobutoxy)benzoate

  • MDL number MFCD08447794
  • Cas Number : 124064-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130342
Availability details for
Quantity
  • Availability
  • Add to Cart