ENA394768184 Enamine

1-chloroethyl propyl carbonate

  • MDL number MFCD20624698
  • Cas Number : 99464-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130361
Availability details for
Quantity
  • Availability
  • Add to Cart