ENA394768185 Enamine

1-[(tert-butoxy)carbonyl]-2-methylpiperidine-2-carboxylic acid

  • MDL number MFCD09910215
  • Cas Number : 746658-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129692
Availability details for
Quantity
  • Availability
  • Add to Cart