ENA394768187 Enamine

methyl azepane-2-carboxylate hydrochloride

  • MDL number MFCD16660246
  • Cas Number : 34459-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130367
Availability details for
Quantity
  • Availability
  • Add to Cart