ENA394768188 Enamine

tert-butyl pyridine-2-carboxylate

  • MDL number MFCD19600064
  • Cas Number : 79233-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130368
Availability details for
Quantity
  • Availability
  • Add to Cart