ENA394768191 Enamine

1-(chloromethyl)-4-(propan-2-ylsulfanyl)benzene

  • MDL number MFCD21911666
  • Cas Number : 1368614-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130384

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130384
Availability details for
Quantity
  • Availability
  • Add to Cart