ENA394768192 Enamine

2-[4-(propan-2-ylsulfanyl)phenyl]acetic acid

  • MDL number MFCD00837332
  • Cas Number : 3583-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130385
Availability details for
Quantity
  • Availability
  • Add to Cart