ENA394768194 Enamine

4-(1,2-oxazol-5-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD19200228
  • Cas Number : 5765-41-3
  • Molecular Weight : 243.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 147-149
Iupac Name 4-(5-isoxazolyl)benzenesulfonyl chloride
Inchi Code 1S/C9H6ClNO3S/c10-15(12,13)8-3-1-7(2-4-8)9-5-6-11-14-9/h1-6H
Inchi Key YLMBEKRVKFZJCC-UHFFFAOYSA-N

Description

Melting Point 147-149
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130389

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-130389
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart