ENA394768196 Enamine

methyl 3-[(2,2-dimethoxyethyl)amino]propanoate

  • MDL number MFCD16620550
  • Cas Number : 86017-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130394
Availability details for
Quantity
  • Availability
  • Add to Cart