ENA394768198 Enamine

tert-butyl N-{[4-amino-5-({[(tert-butoxy)carbonyl]amino}methyl)-4H-1,2,4-triazol-3-yl]methyl}carbamate

  • MDL number MFCD17779777
  • Cas Number : 1253420-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130428
Availability details for
Quantity
  • Availability
  • Add to Cart