ENA394768199 Enamine

4,4-dimethyl-2-[(trimethylsilyl)oxy]pentanenitrile

  • MDL number MFCD17254851
  • Cas Number : 1344264-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130459
Availability details for
Quantity
  • Availability
  • Add to Cart