ENA394768201 Enamine

3-(2-bromophenyl)-1-(pyridin-4-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD13724487
  • Cas Number : 1155571-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129698
Availability details for
Quantity
  • Availability
  • Add to Cart