ENA394768204 Enamine

5-(pyridin-2-yl)pyridine-3-carboxylic acid

  • MDL number MFCD18855387
  • Cas Number : 35989-05-0
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2,3'-bipyridine]-5'-carboxylic acid
Inchi Code 1S/C11H8N2O2/c14-11(15)9-5-8(6-12-7-9)10-3-1-2-4-13-10/h1-7H,(H,14,15)
Inchi Key CETDUWNFRJWXRN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130503
Availability details for
Quantity
  • Availability
  • Add to Cart