ENA394768210 Enamine

2-[2-(trifluoromethyl)phenyl]morpholine

  • MDL number MFCD11646231
  • Cas Number : 1094649-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130512

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130512
Availability details for
Quantity
  • Availability
  • Add to Cart