ENA394768214 Enamine

2-(3,4-dichlorophenyl)-5-methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07379223
  • Cas Number : 1443979-88-1
  • Molecular Weight : 272.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,4-dichlorophenyl)-5-methyl-1,3-oxazole-4-carboxylic acid
Inchi Code 1S/C11H7Cl2NO3/c1-5-9(11(15)16)14-10(17-5)6-2-3-7(12)8(13)4-6/h2-4H,1H3,(H,15,16)
Inchi Key ULCCLFXRXPQHJE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130531
Availability details for
Quantity
  • Availability
  • Add to Cart