ENA394768218 Enamine

6-chloro-1H-1,3-benzodiazol-4-amine

  • MDL number MFCD18807327
  • Cas Number : 10597-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130536
Availability details for
Quantity
  • Availability
  • Add to Cart