ENA394768221 Enamine

3-(3-aminophenyl)propan-1-ol

  • MDL number MFCD09927367
  • Cas Number : 52273-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129707
Availability details for
Quantity
  • Availability
  • Add to Cart