ENA394768225 Enamine

1-(piperidin-1-yl)prop-2-en-1-one

  • MDL number MFCD00085224
  • Cas Number : 10043-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130545

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130545
Availability details for
Quantity
  • Availability
  • Add to Cart