ENA394768226 Enamine

1-(2,4-difluorophenyl)cyclopentane-1-carbonitrile

  • MDL number MFCD11036735
  • Cas Number : 1260758-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130557
Availability details for
Quantity
  • Availability
  • Add to Cart