ENA394768228 Enamine

1-methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD12025862
  • Cas Number : 130530-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130560

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130560
Availability details for
Quantity
  • Availability
  • Add to Cart