ENA394768232 Enamine

3-methyl-1-(6-methylpyridazin-3-yl)-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD15492106
  • Cas Number : 1443979-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130567
Availability details for
Quantity
  • Availability
  • Add to Cart