ENA394768235 Enamine

3-chloropyridine-4-carboxamide

  • MDL number MFCD09907954
  • Cas Number : 100859-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129494
Availability details for
Quantity
  • Availability
  • Add to Cart