ENA394768237 Enamine

3-chloro-N-[(4-chlorophenyl)methyl]aniline

  • MDL number MFCD11116133
  • Cas Number : 107623-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130595
Availability details for
Quantity
  • Availability
  • Add to Cart