ENA394768242 Enamine

1-{[2-(aminomethyl)phenyl]methyl}pyrrolidin-3-ol

  • MDL number MFCD17249249
  • Cas Number : 1340306-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130613
Availability details for
Quantity
  • Availability
  • Add to Cart