ENA394768249 Enamine

3-(1,2-oxazol-5-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD19200322
  • Cas Number : 13567-19-6
  • Molecular Weight : 243.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(5-isoxazolyl)benzenesulfonyl chloride
Inchi Code 1S/C9H6ClNO3S/c10-15(12,13)8-3-1-2-7(6-8)9-4-5-11-14-9/h1-6H
Inchi Key IMCKOKLNTSTPBX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130654
Availability details for
Quantity
  • Availability
  • Add to Cart