ENA394768250 Enamine

6-carbamoylpyridine-2-carboxylic acid

  • MDL number MFCD12022370
  • Cas Number : 97310-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130656
Availability details for
Quantity
  • Availability
  • Add to Cart