ENA394768254 Enamine

3-methyl-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD18432182
  • Cas Number : 24454-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130667
Availability details for
Quantity
  • Availability
  • Add to Cart