ENA394768255 Enamine

3-bromo-2-hydroxy-5-methylbenzaldehyde

  • MDL number MFCD00275688
  • Cas Number : 33172-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129716
Availability details for
Quantity
  • Availability
  • Add to Cart