ENA394768257 Enamine

4-(benzylamino)butanenitrile hydrochloride

  • MDL number MFCD24843069
  • Cas Number : 7544-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130693
Availability details for
Quantity
  • Availability
  • Add to Cart