ENA394768259 Enamine

2-(5-fluoro-2-methylphenyl)-2-oxoacetic acid

  • MDL number MFCD07698685
  • Cas Number : 890097-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130753
Availability details for
Quantity
  • Availability
  • Add to Cart