ENA394768261 Enamine

1-[(4-cyanophenyl)carbamoyl]piperidine-2-carboxylic acid

  • MDL number MFCD09729885
  • Cas Number : 1103815-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130767
Availability details for
Quantity
  • Availability
  • Add to Cart