ENA394768266 Enamine

N-[(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methylidene]hydroxylamine

  • MDL number MFCD00121970
  • Cas Number : 27006-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129739

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129739
Availability details for
Quantity
  • Availability
  • Add to Cart