ENA394768267 Enamine

methyl 3-(3,4-dimethylphenyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD13967203
  • Cas Number : 192701-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130888
Availability details for
Quantity
  • Availability
  • Add to Cart