ENA394768278 Enamine

N-(2-hydroxyethyl)pyridine-4-carboxamide

  • MDL number MFCD00075311
  • Cas Number : 6265-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15591
Availability details for
Quantity
  • Availability
  • Add to Cart