ENA394768280 Enamine

1-[(3,4-dimethoxybenzene)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD03719881
  • Cas Number : 593261-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15803
Availability details for
Quantity
  • Availability
  • Add to Cart