ENA394768285 Enamine

2-(prop-2-en-1-ylsulfanyl)-1H-1,3-benzodiazole

  • MDL number MFCD00124393
  • Cas Number : 51389-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-16177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16177
Availability details for
Quantity
  • Availability
  • Add to Cart