ENA394768319 Enamine

N-(4-hydroxyphenyl)-3,4,5-trimethoxybenzamide

  • MDL number MFCD00705659
  • Cas Number : 15958-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17105
Availability details for
Quantity
  • Availability
  • Add to Cart