ENA394768343 Enamine

3-bromo-4-(4-fluorophenyl)-4-oxobutanoic acid

  • MDL number MFCD06335536
  • Cas Number : 49780-08-7
  • Molecular Weight : 275.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-17687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17687
Availability details for
Quantity
  • Availability
  • Add to Cart