ENA394768384 Enamine

3-[(4-fluorophenyl)sulfanyl]propanoic acid

  • MDL number MFCD00236613
  • Cas Number : 19543-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18770

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18770
Availability details for
Quantity
  • Availability
  • Add to Cart