ENA394768387 Enamine

potassium 3-benzyl-7-ethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-ide

  • MDL number MFCD06671610
  • Cas Number : 1052544-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18789

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18789
Availability details for
Quantity
  • Availability
  • Add to Cart