ENA394768456 Enamine

3,4-dichloro-5-hydroxy-2,5-dihydrofuran-2-one

  • MDL number MFCD00272415
  • Cas Number : 766-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...182
Purity 95.00
Flash Point 186
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21576

Safety Information

signalword Danger
safety_statement 26-36/37/39-45
msds https://www.enaminestore.com/catalog/EN300-21576
pictograms GHS05,GHS06,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart