ENA394768465 Enamine

1-[(4-fluorobenzene)sulfonyl]pyrrolidine

  • MDL number MFCD00783756
  • Cas Number : 157187-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24092
Availability details for
Quantity
  • Availability
  • Add to Cart