ENA394768466 Enamine

4-(4-fluorobenzenesulfonyl)morpholine

  • MDL number MFCD00194713
  • Cas Number : 383-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24120
Availability details for
Quantity
  • Availability
  • Add to Cart