ENA394768472 Enamine

N-{4-[(2-chloroacetamido)methyl]phenyl}cyclopropanecarboxamide

  • MDL number MFCD11547007
  • Cas Number : 1098346-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28311
Availability details for
Quantity
  • Availability
  • Add to Cart